Learn More
SP Bel-Art™ H-B Durac Plus Oven Verification Thermometer
Shop All SP Scienceware Products
Click to view available options
Temperature Range (Metric):
20°C to 130°C
50°C to 200°C
Unit Size:
Each
Description
- Verify sample temperatures in ovens
- Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
- Insert the thermometer into the bottle and place near the samples for best results
- Thermometers contain organic liquid, or EnviroKleen™ certified Enviro-Safe™ liquid, and feature permanently fused markings and lead free glass
- PFA coating on select thermometers safely contain glass and liquid in case of breakage
- Manufactured and tested by our USA quality team; individually serialized
- Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available
Specifications
Specifications
| Bottle Fill | Glass Beads |
| Bottle Size | 30 mL |
| Length (Metric) | 145 mm |
| Thermometer Fill | Organic Liquid |
| Product Type | Thermometer |
| Length (English) | 5.7 in. |
| Temperature Range (Metric) | 50°C to 200°C |
| Accuracy | ±2.5°C; ±5°C above 100°C |