missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Gypsy Moth Diet Without Agar
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
Description
- Available with agar included, packaged separately or omitted completely
- Methyl paraben and sorbic acid added as preservatives
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Product Type | Animal Diet |
| Disposable | Single-use |
| Quantity | 1 L |
| Sterility | Non-sterile |
| Storage Requirements | Room Temperature |
| Length (English) | 9 in. |
| Length (Metric) | 0.2286 m |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction