Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ Column
Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
1 x 1 mL
1 x 5 mL
5 x 0.2 mL
1 x 10 mL
Quantity:
1 Column
5 Columns
Unit Size:
Each
Pack of 5
Description
- Strep-Tactin™XT Superflow™ columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which leads to an outstanding purity of the target protein.
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
- In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
- To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Specifications
| Capacity (Metric) | 1 x 1 mL |
| Quantity | 1 Column |
| For Use With (Application) | Protein Purification |
| Type | Gravity Flow Column |
For Research Use Only.