missing translation for 'onlineSavingsMsg'
Learn More

GRAMD1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24905-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893415

  • 4593.76 NOK / 150µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GRAMD1B, also named as KIAA1201, contains a transmembrane region and two domains of known function: the GRAM domain and a VASt domain. It is predicted to localize in the nucleus, supported by several nuclear transport signals and nuclear associated motifs. This highly conserved gene is found in a variety of vertebrates and invertebrates, however is not found in bacteria or fungi. There are some isoforms with MW 50 kDa and 81-87 kDa. With phosphor modification, it is about 90-110 kDa in WB detection.
Specifications

Specifications

GRAMD1B
Polyclonal
Unconjugated
GRAMD1B
GRAM domain containing 1B, GRAMD1B, KIAA1201
Rabbit
Antigen Affinity Chromatography
RUO
235283, 57476
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3KR37, Q80TI0
GRAMD1B
GRAMD1B Fusion Protein Ag20670
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers