missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAMD1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | GRAMD1B |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803425
|
Proteintech
24905-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893415
|
Proteintech
24905-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
GRAMD1B, also named as KIAA1201, contains a transmembrane region and two domains of known function: the GRAM domain and a VASt domain. It is predicted to localize in the nucleus, supported by several nuclear transport signals and nuclear associated motifs. This highly conserved gene is found in a variety of vertebrates and invertebrates, however is not found in bacteria or fungi. There are some isoforms with MW 50 kDa and 81-87 kDa. With phosphor modification, it is about 90-110 kDa in WB detection.Specifications
GRAMD1B | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q3KR37, Q80TI0 | |
235283, 57476 | |
GRAMD1B Fusion Protein Ag20670 | |
Primary | |
-20°C | |
GRAMD1B |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GRAM domain containing 1B, GRAMD1B, KIAA1201 | |
GRAMD1B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |