Learn More
- Home
- Products
- Extraction Systems
- Extraction Thimbles
- Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble
Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble
Product Code. 11724053 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inside:
16 mm
19 mm
22 mm
25 mm
26 mm
27 mm
28 mm
30 mm
33 mm
34 mm
35 mm
37 mm
41 mm
43 mm
58 mm
70 mm
80 mm
Length (Metric):
40 mm
60 mm
80 mm
90 mm
94 mm
100 mm
118 mm
120 mm
123 mm
130 mm
150 mm
170 mm
200 mm
240 mm
250 mm
Unit Size:
Pack of 25
29 product options available for selection
| Product Code. | Diameter (Metric) Inside | Length (Metric) | Unit Size |
|---|---|---|---|
| 11724053 | 33 mm | 80 mm | Pack of 25 |
| 11764526 | 16 mm | 100 mm | Pack of 25 |
| 11734536 | 19 mm | 40 mm | Pack of 25 |
| 11724043 | 19 mm | 90 mm | Pack of 25 |
| 11774526 | 22 mm | 60 mm | Pack of 25 |
| 11754043 | 25 mm | 100 mm | Pack of 25 |
| 11734043 | 22 mm | 80 mm | Pack of 25 |
| 11774043 | 26 mm | 60 mm | Pack of 25 |
| 11744043 | 25 mm | 80 mm | Pack of 25 |
| 11784526 | 27 mm | 80 mm | Pack of 25 |
| 11794043 | 28 mm | 100 mm | Pack of 25 |
| 11542103 | 28 mm | 120 mm | Pack of 25 |
| 11784043 | 28 mm | 80 mm | Pack of 25 |
| 11714053 | 30 mm | 100 mm | Pack of 25 |
| 11724536 | 30 mm | 150 mm | Pack of 25 |
| 11704053 | 30 mm | 80 mm | Pack of 25 |
| 11734053 | 33 mm | 100 mm | Pack of 25 |
| 11744536 | 33 mm | 118 mm | Pack of 25 |
| 11714536 | 33 mm | 94 mm | Pack of 25 |
| 11794526 | 34 mm | 130 mm | Pack of 25 |
| 11764536 | 35 mm | 150 mm | Pack of 25 |
| 11744053 | 37 mm | 130 mm | Pack of 25 |
| 11754053 | 41 mm | 123 mm | Pack of 25 |
| 11704536 | 41 mm | 150 mm | Pack of 25 |
| 11754536 | 43 mm | 123 mm | Pack of 25 |
| 11774536 | 58 mm | 170 mm | Pack of 25 |
| 11784536 | 70 mm | 240 mm | Pack of 25 |
| 11794536 | 80 mm | 200 mm | Pack of 25 |
| 11704546 | 80 mm | 250 mm | Pack of 25 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
Description
- 100% High pure cotton linters fibers
- Neutral and fat free
- High mechanical strength
- Excellent retention capacity
Applications:
- Extraction of sulphur
- Extraction of the content of free lipids in food
- Determination of dust and tar in industrial air and gases
- Lacquer, binder analysis in paint colours
- Control of emission smokes in industrial chimneys
- Gravimetric analysis of dust in gases
Specifications
Specifications
| Diameter (Metric) Inside | 33 mm |
| Material | High Purity Cellulose Fibers and Cotton Linters |
| For Use With (Application) | Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases |
| Grade | 7 |
| For Use With (Equipment) | Suitable for all extractors systems |
| Length (Metric) | 80 mm |
| Product Type | Extraction Thimble |