missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR101 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20074-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to GPR101.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. Expression of GPR101 has been documented by Northern analysis in human hypothalamus, caudate, and putamen but not in thalamus, hippocampus, frontal cortex, or pons.Specifications
| GPR101 | |
| Polyclonal | |
| Unconjugated | |
| GPR101 | |
| G protein coupled receptor 101, GPCR6, GPR101 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83550 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96P66 | |
| GPR101 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Non-human Primate | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction