Learn More
Trajan Scientific and Medical™ GP GC Septa
Combines significantly longer injection life, low bleed and low injection port adhesion
523.00 NOK - 555.00 NOK
Specifications
Material Silicone
Temperature (Metric) Max 275°C
Quantity 50 Pack
View More Specs
Products 4
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
10370991
View Documents Promotion Details
Trajan Scientific and Medical™
041821
6 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10646031
View Documents Promotion Details
Trajan Scientific and Medical™
041823
8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10248910
View Documents Promotion Details
Trajan Scientific and Medical™
041822
7 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10441391
View Documents Promotion Details
Trajan Scientific and Medical™
041824
9 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Designed for non-demanding, routine applications
  • Reliable seal against carrier gas pressure in inlet
  • No bleed or contamination into GC system
  • Maximum operating temperature of 275°C
Specifications
Specifications
Silicone
275°C
50 Pack