missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ GP GC Septa

Combines significantly longer injection life, low bleed and low injection port adhesion

Brand:  Trajan Scientific and Medical™ 041822

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10248910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Designed for non-demanding, routine applications
  • Reliable seal against carrier gas pressure in inlet
  • No bleed or contamination into GC system
  • Maximum operating temperature of 275°C
Specifications

Specifications

7 mm
275°C
50 Pack
Silicone
GP Septa
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers