missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOSR2/Membrin Mouse anti-Human, Clone: 7E7C7, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | GOSR2/Membrin |
|---|---|
| Clone | 7E7C7 |
| Concentration | 1.11 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857735
|
Proteintech
66134-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847735
|
Proteintech
66134-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.Specifications
| GOSR2/Membrin | |
| 1.11 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 27 kDa Golgi SNARE protein, Bos1, GOSR2, GS27, Membrin | |
| GOSR2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 7E7C7 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O14653 | |
| 9570 | |
| GOSR2/Membrin Fusion Protein Ag21393 | |
| Primary | |
| -20°C | |
| GOSR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title