missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOSR2/Membrin Mouse anti-Human, Clone: 7E7C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66134-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.Specifications
| GOSR2/Membrin | |
| Monoclonal | |
| 1.11 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14653 | |
| GOSR2 | |
| GOSR2/Membrin Fusion Protein Ag21393 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 7E7C7 | |
| Unconjugated | |
| GOSR2 | |
| 27 kDa Golgi SNARE protein, Bos1, GOSR2, GS27, Membrin | |
| Mouse | |
| Protein A | |
| RUO | |
| 9570 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction