missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Goat anti-Rabbit IgG, Whole Molecule, Affinity Gel

Product Code. 11450095 Shop All MP Biomedicals Products
Click to view available options
:
Goat anti-Rabbit IgG, Whole Molecule, Affinity Gel; Quantity: 2 mL
This item is not returnable. View return policy

Product Code. 11450095

Brand: MP Biomedicals™ 0855696

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Goat anti-Rabbit IgG, Whole Molecule, Affinity Gel is a goat affinity purified antibody to rabbit IgG (whole molecule) covalently coupled to 2 mL of Sepharose-4B, and packed in a polystyrene chromatography column.

  • Host Species: Goat
  • Species Reactivity: Rb
  • Antibody Purification
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mL
Label or Dye Unlabeled
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.