missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL3 Monoclonal Antibody (OTI5B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813294S
Additional Details : Weight : 0.05000kg
Description
GNL3 Monoclonal antibody specifically detects GNL3 in Human samples. It is validated for Western BlotSpecifications
GNL3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BVP2 | |
Mouse | |
Protein A/G | |
RUO | |
26354 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI5B1 | |
Unconjugated | |
GNL3 | |
C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin | |
Full length human recombinant protein of human GNL3 (NP_055181) produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |