missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL3 Monoclonal Antibody (OTI5B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2816.72 NOK - 6295.38 NOK
Specifications
| Antigen | GNL3 |
|---|---|
| Clone | OTI5B1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17598124
|
Origene Technologies
TA813294S |
30 μL |
2816.72 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17578124
|
Origene Technologies
TA813294 |
100 μL |
6295.38 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GNL3 Monoclonal antibody specifically detects GNL3 in Human samples. It is validated for Western BlotSpecifications
| GNL3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin | |
| Full length human recombinant protein of human GNL3 (NP_055181) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| GNL3 |
| OTI5B1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BVP2 | |
| 26354 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title