missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GNGT1 (Human) Recombinant Protein
Human GNGT1 full-length ORF ( AAH29367, 1 a.a. - 74 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002792-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPVINIEDLTEKDKLKMEVDQLKKEVTLERMLVSKCCEEVRDYVEERSGEDPLVKGIPEDKNPFKELKGGCVIS
Specifications
AAH29367 | |
guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNGT1 | |
GST |
2792 | |
Wheat germ expression system | |
25 μg | |
GNG1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |