Learn More
GMDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15442-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the conversion of GDP-D-mannose to GDP-4-dehydro-6-deoxy-D-mannose.Specifications
| GMDS | |
| Polyclonal | |
| Unconjugated | |
| GMDS | |
| GDP D mannose dehydratase, GDP mannose 4,6 dehydratase, GMD, GMDS, SDR3E1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 218138, 2762, 291095 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60547, Q8K0C9 | |
| GMDS | |
| GMDS Fusion Protein Ag7319 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |