missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
1575.00 NOK - 5045.00 NOK
Specifications
| Antigen | Glut1 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240463
|
Novus Biologicals
NB120-15309-0.1ML |
0.1 mL |
1575.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18720784
|
Novus Biologicals
NB120-15309 |
5045.00 NOK
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Glut1 Polyclonal specifically detects Glut1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Glut1 | |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| P11166 | |
| SLC2A1 | |
| IgG | |
| Affinity purified | |
| This antibody reacts with a 55 kDa protein. Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response in insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus and breast. GLUT-1 immunoreactivity in some cancers, including trans-carcinoma of the urinary bladder, has been associated with aggressive behavior. |
| 0.2 mg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| 6513 | |
| A synthetic peptide derived from the C-terminal region of human GLUT1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title