missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-15309-0.1ML
This item is not returnable.
View return policy
Description
Glut1 Polyclonal specifically detects Glut1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Glut1 | |
| Polyclonal | |
| Unconjugated | |
| Citrate, pH 6.0, containing less than 0.1% sodium azide with Sodium Azide | |
| DYT18DYT17, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, GLUT1GLUT, HepG2 glucose transporter, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| This antibody reacts with a 55 kDa protein. Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response in insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus and breast. GLUT-1 immunoreactivity in some cancers, including trans-carcinoma of the urinary bladder, has been associated with aggressive behavior. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| 0.2 mg/mL | |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P11166 | |
| SLC2A1 | |
| A synthetic peptide derived from the C-terminal region of human GLUT1. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 6513 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction