missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLT25D2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25993-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
GLT25D2 has a beta-galactosyltransferase activity and transfers beta-galactose to hydroxylysine residues on collagen.Specifications
GLT25D2 | |
Polyclonal | |
Unconjugated | |
COLGALT2 | |
C1orf17, GLT25D2, KIAA0584 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23127, 269132 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NVG7, Q8IYK4 | |
COLGALT2 | |
GLT25D2 Fusion Protein Ag23360 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |