missing translation for 'onlineSavingsMsg'
Learn More

GLT25D2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen GLT25D2
Concentration 0.21 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16804525
View Documents Promotion Details
Proteintech
25993-1-AP-20UL
20 μL
1611.84 NOK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16894515
View Documents Promotion Details
Proteintech
25993-1-AP-150UL
150 μL
4593.76 NOK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GLT25D2 has a beta-galactosyltransferase activity and transfers beta-galactose to hydroxylysine residues on collagen.
Specifications

Specifications

GLT25D2
Western Blot
Unconjugated
Rabbit
Human, Mouse
Q6NVG7, Q8IYK4
23127, 269132
GLT25D2 Fusion Protein Ag23360
Primary
-20°C
COLGALT2
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
C1orf17, GLT25D2, KIAA0584
COLGALT2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications