Learn More
STEMCELL Technologies GloCell™ Fixable Viability Dye Red 780
Click to view available options
Quantity:
100 Tests
5 x 100 Tests
Unit Size:
100 tests
Pack of 5
Description
- GloCell™ Fixable Viability Dye Red 780 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
- The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation
- Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay
- GloCell™ Fixable Viability Dye Red 780 is excited by the red laser at 633 nm and it has a fluorescence emission maximum of 780 nm that can be detected using the 780/60 band pass filter used to detect APC-Cy7
Specifications
Specifications
| For Use With (Application) | Flow Cytometry; Immunofluorescence |
| Quantity | 100 Tests |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.