Learn More
Fisherbrand™ Glass Screw Neck Specimen Vials
Product Code. p-7070786 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1.75 mL
3.5 mL
7 mL
10.5 mL
14 mL
21 mL
28 mL
Closure Size:
10 mm
15 mm
18 mm
22 mm
28 mm
Style:
Squat form
Tall form
Unit Size:
Pack of 140
Pack of 190
Pack of 231
Pack of 264
Pack of 399
Pack of 666
Pack of 882
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Closure Size Style Unit Size
15364779 28 mL 28 mm Squat form Pack of 140
15394779 7 mL 15 mm Tall form Pack of 399
15304789 10.5 mL 15 mm Tall form Pack of 399
15354779 14 mL 22 mm Squat form Pack of 190
15314789 14 mL 18 mm Tall form Pack of 264
15324789 21 mL 18 mm Tall form Pack of 231
15344779 7 mL 18 mm Squat form Pack of 264
15374779 1.75 mL 10 mm Tall form Pack of 882
15384779 3.5 mL 10 mm Tall form Pack of 666
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 15364779
Add to basket
Edge
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Store specimens securely with squat or tall form glass vials. Fisherbrand™ Glass Screw Neck Specimen Vials are available in a variety of sizes. Fisherbrand™ polypropylene screw caps are sold separately.
  • Manufactured from clear Type 1B neutral glass
  • Squat and tall form options with standard neck sizes
  • 1 dram equates to approximately 3.55mL (or 0.125 fl. oz.)
  • Suitable for media, diagnostic, storage, display, and sample collection applications
  • Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials sold separately
Specifications
Color Clear
Diameter (Metric) Outer 27 mm
Material Neutral Glass (Type 1B)
Volume (Metric) 28 mL
Style Squat form
Closure Type Screw Cap
Closure Size 28 mm
Height (Metric) 72 mm