Learn More
Fisherbrand™ Glass Bottom for Desiccator
Product Code. p-7083455 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Outer:
94 mm
143 mm
192 mm
239 mm
285 mm
Height (Metric):
113 mm
158 mm
203 mm
238 mm
288 mm
For Use With (Equipment):
Desiccator (Type: 100mm)
Desiccator (Type: 150mm)
Desiccator (Type: 200mm)
Desiccator (Type: 250mm)
Desiccator (Type: 300mm)
Unit Size:
Each
Product Code. Diameter (Metric) Outer Height (Metric) For Use With (Equipment) unitSize
15554635 239 mm 238 mm Desiccator (Type: 250mm) Each
15554625 143 mm 158 mm Desiccator (Type: 150mm) Each
15504645 285 mm 288 mm Desiccator (Type: 300mm) Each
15504635 192 mm 203 mm Desiccator (Type: 200mm) Each
15504625 94 mm 113 mm Desiccator (Type: 100mm) Each
5 options
Product Code. 15554635
Only null left
Add to basket
Add to basket
Glass only bottom for use with desiccator
  • Bottom for desiccator
Certifications
ISO 13 130, DIN 12 491
Specifications
Diameter (Metric) Outer 239 mm
Product Type Bottom
Description I.D.: 131 mm
Material Glass
For Use With (Equipment) Desiccator (Type: 250mm)
Height (Metric) 238 mm