Learn More
Fisherbrand™ Glass Body pH Electrode, S7
Product Code. 11786338 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
160 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11786338 160 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11786338
  • ONLINE EXCLUSIVE PRICE
    946.00 NOK
    835.71 NOK / Pack of 1
    Save 110.29 NOK
    11% Off
View all promotions
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
General purpose, standard, glass body pH Meter with an S7 connector. Fisherbrand™ Glass Body pH Electrode, S7 uses a KCl 4M + AGCl electrolyte.
  • General purpose
  • Glass body
  • pH range: 0 - 14
  • Electrolyte: KCl 4M + AgCl
  • Connector: S7
Specifications
Connector Types S7
Diameter (Metric) 12 mm
Length (Metric) 160 mm
pH Range 0 to 14
Temperature (Metric) Range 0°C to 80°C