missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GKN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24651-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
GKN2 | |
Polyclonal | |
Unconjugated | |
GKN2 | |
BLOT, Blottin, gastrokine 2, GDDR, GKN2, PRO813, TFIZ1, VLTI465 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
200504, 66284 | |
-20°C | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86XP6, Q9CQS6 | |
Gkn2 | |
GKN2 Fusion Protein Ag19153 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |