missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GKN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | GKN2 |
---|---|
Concentration | 0.6 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803145
|
Proteintech
24651-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893135
|
Proteintech
24651-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The function of this protein remains unknown.Specifications
GKN2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q86XP6, Q9CQS6 | |
200504, 66284 | |
GKN2 Fusion Protein Ag19153 | |
Primary | |
-20°C | |
GKN2 |
0.6 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BLOT, Blottin, gastrokine 2, GDDR, GKN2, PRO813, TFIZ1, VLTI465 | |
Gkn2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |