Description
Universal Multichannel Stand is made of AltuglasTM Essential quality PMMA (Polymethyl Methacrylate). PMMA is highly transparent and offers a good resistance to scratches, impacts and temperature. It also has a good resistance to many chemicals commonly handled in laboratories such as alcohols like ethanol and methanol, disinfectants and some acids and bases depending on the concentration.
- Made of flexible plastic
- Support edges widen or retract to adapt geometry of virtually all pipet brands
Specifications
Specifications
| Content And Storage | 1x Multichannel pipette stand |
| Color | Clear |
| For Use With | Pipettes |
| Material | PMMA |
| Type | Pipette Holder |
| For Use With (Equipment) | All Gilson and other brands of multichannel pipet |
| Includes | 1x Multichannel pipette stand |
| Packaging Type | Box |
| Quantity | 1 |
| Dimensions (L x W x H) | 151.9 X 151.9 X 105.7 mm |
| Show More |