Learn More
Gilson™ TRACKMANTM Duo-plate Pipeting Tracker
Product Code. 12356172 Shop All Gilson Products
Click to view available options
:
TRACKMANTM Duo-plate Pipeting Tracker; For use with: 96 and 384 Well Microplates, Single and Multichannel Pipets
Product Code. 12356172
Brand: Gilson™ F70301
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
TRACKMANTM Duo-plate Pipeting Tracker with a small footprint will increase productivity and eliminate potential errors.
  • Easily switch from using one size plate to another on the same instrument
  • Timer function enables hands-free operation and optional foot pedal is available for hands-free control
  • Long lasting battery allows cordless operation for up to eight hours so you can take your TRACKMAN anywhere
  • Duo-plate tracker lights the way for pipetting to avoid losing track when pipetting expensive samples
  • Can be used with both 96 and 384 well microplates, and single and multichannel pipettes
TRUSTED_SUSTAINABILITY
Specifications
For Use With All Pipettes with Tips Mounted
Material Polypropylene
Number of Channels 16
Type Foot Pedal
For Use With (Equipment) 96 and 384 Well microplates, single and multichannel Pipette
Includes Long lasting battery
Quantity 1