Learn More
Gilson™ PIPETMANTM TIPACKTM Racked Pipet Tips
GREENER_CHOICE
Product Code. 12649581 Shop All Gilson Products
Click to view available options
Length (Metric):
14.5 cm
32.2 cm
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
100 to 1000 μL
100 to 1200 μL
2 to 200 μL
20 to 300 μL
500 to 5000 μL
Model:
D10
D1000
D1200
D200
D300
D5000
DL10
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 600
Pack of 960
Product Code. 12649581
Brand: Gilson™ F171300
Only null left
Add to basket
Add to basket
Engineered to be functional, convenient and robust. GilsonTM PIPETMANTM TIPACKTM Racked Pipet Tips are available in sterilized or autoclavable packages.
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Requires low tip-mounting force.
  • Robust and designed to withstand autoclaving.
  • Sterility assurance level SAL 10-6 guaranteed, using gamma irradiation.
  • All packs are wrapped in medical device quality plastic which provides a complete shield against contamination.
  • Indicator dots used on Gilson products undergo an irreversible color change upon sterilization; yellow gamma dots turn red when products have been properly exposed.
  • TIPACKs are securely sealed with a sticker; a full product description is given on the TIPACK for traceability and easy identification.
  • Available in sterilized or autoclavable packages.
  • Manufactured in a clean room and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging to meet GLP requirements.
Order Info
10 TIPACKs of 96 racked tips
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 2 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model D200
Length (English) 5.71 in.
Length (Metric) 14.5 cm
Format Rack
Tip Style Standard
Quantity 960 (10x96)
Product Type Pipet Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Pipetman Tipack
Material Polypropylene
Graduations 20 μL; 100 μL