Learn More
- Home
- Products
- Pipette Products
- Pipettes
- Electronic Multichannel Pipettes
- Gilson™ PIPETMAN™ M Pipettes
Gilson™ PIPETMAN™ M Pipettes
Product Code. 15510510 Shop All Gilson Products| Product Code. | Number of Channels | Model | Volume (Metric) | Compatible Tips | unitSize |
|---|---|---|---|---|---|
| 15510510 | 12 | P12x200M | 20 to 200 μL | D200, D300, DF100ST, DF200ST, DF300ST tips | Each |
| 15560500 | 8 | P8x10M | 0.5 to 10 μL | D10, DL10, DF10, DFL10 tips | Each |
| 15570500 | 12 | P12X10M | 0.5 to 10 μL | D10, DL10, DF10 or DFL10 tips | Each |
| 15580500 | 8 | P8X20M | 1 to 20 μL | DL10, DF30ST, D200 tips | Each |
| 15590500 | 12 | P12x20M | 1 to 20 μL | DL10, DF30ST, D200 tips | Each |
| 15500510 | 8 | P8x200M | 20 to 200 μL | D200, D300, DF100ST, DF200ST, DF300ST tips | Each |
| 15520510 | 8 | P8x300M | 10 to 300 μL | D10, DL10, DF10, DFL10 tips | Each |
| 15530510 | 12 | P12x300M | 10 to 300 μL | D10, DL10, DF10 or DFL10 tips | Each |
| 15550490 | 1 | P300M | 20 to 300 μL | D200, DF200, D300, DF300 | Each |
| 15560490 | 1 | P1200M | 100 to 1200 μL | D1000, DF1000, D1200, DF1200 | Each |
| 15570490 | 1 | P5000M | 500 to 5000 μL | D5000 | Each |
| 15580490 | 1 | P10mLM | 1 to 10 mL | D10 mL | Each |
| 15590490 | 8 | P8x100M | 10 to 10 μL | D200 | Each |
| 15500500 | 12 | P12x100M | 10 to 100 μL | DF100 | Each |
| 15510500 | 8 | P8x1200M | 50 to 1200 μL | D1200 | Each |
| 15520500 | 12 | P12x1200M | 50 to 1200 μL | DF1200 | Each |
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
- Motorized piston with adjustable piston speed that is compatible with varying solution densities
- Lockable volume to prevent inaccuracies
- Color-coded push-button corresponds to matching tip packs
- Service alert reminders
- Unique tip holder design allows for a secure fit of most brands of tips
- Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
- Pipette can be used while charging eliminating bottlenecks and downtime
- Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
- Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
- Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Specifications
Specifications
| Compatible Tips | D200, D300, DF100ST, DF200ST, DF300ST tips |
| Color | Yellow |
| Model | P12x200M |
| Number of Channels | 12 |
| Product Line | Pipetman |
| Type | Pipette |
| Volume (Metric) | 20 to 200 μL |
| Finger Style | Ergonomic |
| Warranty | 1 Year |