Learn More
Gilson™ PIPETMANTM EXPERTTM Universal Fit Gel Loading Pipette Tips: Nonsterile
Shop All Gilson Products
Click to view available options
Tip Style:
Flat, Gel Loading
Gel Loading, Round
Graduations:
2μL
5μL
Unit Size:
Pack of 800
Description
- Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
- Features graduations at varying volumes allowing for quick-checks when aspirating samples
- Sterile model also available
Specifications
Specifications
| Volume (Metric) | 1 to 200 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Model | Gel Loading |
| Length (English) | 12.75 in. |
| Length (Metric) | 8.255cm |
| Clean Claims | RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free |
| For Use With (Application) | Gel Loading |
| Show More |