Learn More
Gilson™ MICROMAN™ Positive-Displacement Pipets
Shop All Gilson Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 to 10 μL
10 to 100 μL
100 to 1000 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
M10
M100
M1000
M25
M250
M50
Compatible Tips:
C10
C100
C1000
C25
C250
C50
missing translation for 'unitSize'
Each
Description
- Piston is located inside the tip or capillary preventing pipet contamination
- Color-coded push-button allows for easy identification of different models
- Dual function push-button that also ejects capillary pistons
- Comfortable PVDF handle resistant to aggressive chemicals
- Accurately measure high vapor pressure liquids without leakage or degradation of the tip
- M10 and M100 models are fully autoclavable
Specifications
Specifications
| Compatible Tips | C1000 |
| Autoclavable | Autoclavable |
| Model | M1000 |
| Number of Channels | 1 |
| Type | Positive Displacement Pipette |
| Volume (Metric) | 100 to 1000 μL |
| For Use With (Application) | Pipetting of viscous and dense liquids |