Learn More
- Home
- Products
- Pipette Products
- Pipette Tips
- Specialty Pipette Tips
- Gilson™ MICROMANTM E Capillary Pistons
Gilson™ MICROMANTM E Capillary Pistons
Product Code. 10789324
Shop All Gilson Products
| Product Code. | Quantity | Volume (Metric) | Model | Format | unitSize |
|---|---|---|---|---|---|
| 10789324 | 192 Pk. | 10 to 100 μL | CP100ST | Box | Pack of 192 |
| 10192852 | 192 Pk. | 1 to 10 μL | CP10ST | Rack, 2 x 96 | Pack of 192 |
| 10011912 | 960 Pk. | 1 to 10 μL | CP10ST | Rack, 10 x 96 | Pack of 960 |
| 15306564 | 576 Pk. | 3 to 25 μL | CP25ST | Box | Pack of 576 |
| 11327920 | 960 Pk. | 10 to 100 μL | CP100ST | Box | Pack of 960 |
| 15304785 | 576 Pk. | 20 to 50 μL | CP50ST | Box | Pack of 576 |
| 15314785 | 576 Pk. | 50 to 250 μL | CP250ST | Box | Pack of 576 |
| 12609621 | 182 Pk. | 100 to 1000 μL | CP1000ST | Box | Pack of 182 |
| 10041932 | 960 Pk. | 1 to 10 μL | CP10 | Rack, 2 x 96 | Pack of 192 |
| 12001532 | 960 Pk. | 1 to 10 μL | CP10 | Rack, 10 x 96 | Pack of 960 |
| 10463942 | 200 Pk. | 3 to 25 μL | CP25 | Box | Pack of 200 |
| 10726470 | 960 Pk. | 3 to 25 μL | CP25 | Rack, 10 x 96 | Pack of 960 |
| 10646653 | 192 Pk. | 10 to 100 μL | CP100 | Box | Pack of 192 |
| 10494132 | 960 Pk. | 10 to 100 μL | CP100 | Box | Pack of 960 |
| 12123771 | 200 Pk. | 20 to 50 μL | CP50 | Box | Pack of 200 |
| 10866510 | 960 Pk. | 20 to 50 μL | CP50 | Rack, 10 x 96 | Pack of 960 |
| 10163522 | 200 Pk. | 50 to 250 μL | CP250 | Box | Pack of 200 |
| 10456925 | 10 x 96 Pk. | 50 to 250 μL | CP250 | Box | Pack of 960 |
| 12597526 | 182 Pk. | 100 to 1000 μL | CP1000 | Box | Pack of 182 |
Product Code. 10789324
Brand: Gilson™ F148415
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
- Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
- Made specifically for GilsonTM MICROMANTM E pipets.
- Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
Specifications
Specifications
| Volume (Metric) | 10 to 100 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Model | CP100ST |
| Length (English) | 5.35 in. |
| Length (Metric) | 41.8 mm |
| Format | Box |
| Tip Style | Positive Displacement |
| Packaging | Box |
| Quantity | 192 Pk. |
| Product Type | Capillary Piston |
| Disposable | Single-use |
| For Use With (Equipment) | Gilson Microman |
| Material | Polypropylene |
| Fit for Purpose | Certified Quality |
| Show More Show Less |