Learn More
Gilson™ MICROMANTM E Capillary Pistons
Shop All Gilson Products
Click to view available options
Quantity:
182 Pk.
192 Pk.
200 Pk.
576 Pk.
960 Pk.
Volume (Metric):
1 to 10 μL
10 to 100 μL
100 to 1000 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
CP10
CP100
CP1000
CP1000ST
CP100ST
CP10ST
CP25
CP250
CP250ST
CP25ST
CP50
CP50ST
Format:
Box
Rack, 10 x 96
Rack, 2 x 96
Unit Size:
Pack of 182
Pack of 192
Pack of 200
Pack of 576
Pack of 960
Description
- Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
- Made specifically for GilsonTM MICROMANTM E pipets.
- Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
Specifications
Specifications
| Volume (Metric) | 50 to 250 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Model | CP250 |
| Length (English) | 4.25 in. |
| Length (Metric) | 86.45 mm |
| Format | Box |
| Tip Style | Positive Displacement |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu