Learn More
Gilson™ Accessory for PIPETMANTM Complete Pipetting System- CarrouselTM Pipet Stand
Product Code. 10571093 Shop All Gilson Products
Change view
Click to view available options
For Use With (Application):
Holds 7 Pipettes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10571093 Holds 7 Pipettes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10571093 Supplier Gilson™ Supplier No. F161401
  • ONLINE EXCLUSIVE PRICE
    1655.00 NOK
    1560.53 NOK / Pack of 1
    Save 94.47 NOK
    5% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
Bulk Quote Request
Convenient rotating stand holds upto seven single and multichannel Gilson pipets
  • Robust, stable and space saving stand
  • Makes bench work look neat and tidy
  • Heavy base of stand prevents toppling over
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1x Carrousel pipette stand
Color White
For Use With Pipettes
Material Polypropylene
Type Pipette Holder
For Use With (Application) Holds 7 Pipettes
For Use With (Equipment) PIPETMAN™ Complete Pipetting System
Includes 1x Carrousel pipette stand
Packaging Type Box
Quantity 1
Dimensions (L x W x H) 205 X 205 X 405 mm
Length (English) 8.1 in
Length (Metric) 205 mm
Width (English) 8.1 in
Width (Metric) 205 mm
Height (English) 15.9 in
Height (Metric) 405 mm
Weight (English) 1.98 lb
Weight (Metric) 780 g