Learn More
Fisherbrand™ PTFE Giant Stir Bar
Product Code. 10410211 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
57 mm
80 mm
108 mm
110 mm
136 mm
159 mm
Diameter (Metric):
14 mm
24 mm
27 mm
36 mm
Shape:
Cylinder
Cylinder, Pivot ring
Triangle
Unit Size:
Each
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Shape unitSize
10410211 57 mm 24 mm Cylinder, Pivot ring Each
10379640 108 mm 24 mm Cylinder, Pivot ring Each
10074400 159 mm 24 mm Cylinder, Pivot ring Each
10166330 108 mm 27 mm Cylinder Each
10369640 159 mm 27 mm Cylinder Each
10470011 57 mm 27 mm Cylinder Each
11868932 80 mm 14 mm Triangle Pack of 5
10480971 80 mm 14 mm Triangle Each
10688071 110 mm 36 mm Triangle Each
12706108 136 mm 36 mm Triangle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Code produit 10410211
Il en reste null
Ajouter au panier
Ajouter au panier
Designed for use in large containers and vials. These giant PTFE-coated magnetic stir bares are autoclavable.
  • Stable, general-purpose magnetic stir bar
  • Designed for use in large containers and vials
  • Autoclavable
Spécification
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Coating PTFE
Diameter (Metric) 24 mm
Length (Metric) 57 mm
Quantity 1/Ea.
Shape Cylinder, Pivot ring
Type Giant PTFE Stir Bar
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.