Includes
Contains12 TAMRA labeled single stranded DNA fragments for use as a standard
The GeneScan™ 500 XL TAMRA™ Size Standard is a TAMRA™ dye-labeled size standard for the reproducible sizing of fragment analysis data.
Order Info
Each
Specifications
Specifications
| Content And Storage | Contains12 TAMRA labeled single stranded DNA fragments for use as a standard. Store the kit at 2-8°C. (Do not freeze). |
| Format | Standard |
| For Use With (Equipment) | 310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Label or Dye | TAMRA™ |
| No. of Reactions | 800 Reactions |
| Product Line | GeneScan, TAMRA |
| Product Type | Dye |
| Shipping Condition | Wet Ice |
| Test Time | Standard |
For research use only