Learn More
Fisherbrand™ Radiation Storage Boxes
Product Code. 12337137 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
60mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12337137 60mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12337137
Estimated Shipment: 15-02-2027
Add to basket
Add to basket
Ideal for protection against Beta or Gamma radiation, these Gamma Midi Duo Radiation boxes with hinged lid and central swivel covers are used in many research, scientific, and other laboratory environments.
  • Gamma Midi Duo 2 Pig-Pen Box 7 x 10 x6cm
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also supplied is range of floor-standing and benchtop bins with anti-slip feet and hinged lids
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
For Use With (Application) To hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Height (Metric) 60mm
Holds Microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Product Type Radiation Storage Box