Learn More
WTW™ Galvanic Dissolved Oxygen Sensors (universal)
Product Code. 10235731
Click to view available options
Length (Metric) Cable:
1.5 m
10 m
15 m
3 m
6 m
Includes:
1 exchange membrane head
3 exchange membrane heads
Unit Size:
Each
Product Code. 10235731
Brand: WTW™ 201547
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get accurate testing results for water and waterous solutions with these membrane-covered galvanic dissolved oxygen sensors that are low maintenance and no polarizarion time required.
  • Dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring
  • Can be used in the lab as well as in the field
  • Features include a quick response time (t99<60 s) and low-maintenance operation
  • Fields of use include surface waters, measurements in wastewater treatment plants, BOD directions
  • Filled with electrolyte and achieves a long dwelling time up to 6 months
  • Calibration vessel is part of the delivery
  • For use with conductivity and multiparameter meters
  • Includes Cable, Cleaning solution, abrasive foil, exchange membrane head(s) and electrolyte
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) Cable 10 m
Includes 3 exchange membrane heads
For Use With (Equipment) Conductivity and multiparameter meters