Learn More
WTW™ Galvanic Dissolved Oxygen Sensor for WTW Conductivity and Multiparameter
Description
The CellOx™ 325 is a dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring, which can be used in the lab as well as in the field. Its features include a quick response time (t99<60 s) and low-maintenance operation. Its fields of use include surface waters, measurements in wastewater treatment plants, BOD directions, but also industrial and other applications. It is filled with electrolyte and achieves a long dwelling time up to 6 months. The calibration vessel is part of the delivery.
Specifications
Specifications
| Description | StirrOx™ G Self-stirring Dissolved Oxygen Sensor |
| Length (Metric) Cable | 20 m |
| Includes | 20m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte |
| For Use With (Equipment) | Conductivity and multiparameter meters |
| Model | StirrOx G |