missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12849-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.Specifications
Galectin 2 | |
Polyclonal | |
Unconjugated | |
LGALS2 | |
Gal 2, Galectin 2, HL14, Lactose binding lectin 2, LGALS2, S Lac lectin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3957 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05162 | |
LGALS2 | |
Galectin 2 Fusion Protein Ag3979 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |