missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Galectin 2 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890784
|
Proteintech
12849-2-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16880784
|
Proteintech
12849-2-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.Specifications
Galectin 2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P05162 | |
3957 | |
Galectin 2 Fusion Protein Ag3979 | |
Primary | |
-20°C | |
LGALS2 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Gal 2, Galectin 2, HL14, Lactose binding lectin 2, LGALS2, S Lac lectin 2 | |
LGALS2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |