Learn More
R&D Systems™ GABA-AR alpha 5 Overexpression Lysate
Product Code. 18125860 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18125860 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18125860 Supplier R&D Systems™ Supplier No. NBP211482
 Restricted Item
Estimated Shipment: 24-11-2026
A Novus product, part of R&D Systems.
The GABA-AR alpha 5 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_115859. The protein contains a C-terminal DDK tag.
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens GABA A Receptor alpha 5, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Product Line Novus
Research Category Cellular Markers, Neuronal Cell Markers, Neurotransmission
Protein State Native
For Use With (Application) Western Blot
For Research Use Only