missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZR1/CDH1 Antibody (AR38.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
1865.00 NOK
Specifications
| Antigen | FZR1/CDH1 |
|---|---|
| Clone | AR38.2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
| Applications | Western Blot, ELISA |
Description
FZR1/CDH1 Monoclonal specifically detects FZR1/CDH1 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
| FZR1/CDH1 | |
| 1.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| anti-CDC20-like protein 1 anti-CDH1 anti-Cdh1/Hct1 homolog anti-Fizzy-related protein homolog anti-FYR anti-Fzr anti-FZR1 anti-hCDH1 CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), FYR, Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242, KIAA1242fizzy-related protein homolog | |
| FZR1 | |
| IgG1 | |
| Protein A purified | |
| NBP2-19018 recognizes Human and mouse. |
| AR38.2 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Mouse | |
| Human, Mouse | |
| 51343 | |
| His-tagged recombinant FZR1/CDH1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title