missing translation for 'onlineSavingsMsg'
Learn More

FZR1/CDH1 Antibody (AR38.2) - BSA Free, Novus Biologicals™

Product Code. 18753284 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
0.03mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18753284 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18753284 Supplier Novus Biologicals Supplier No. NBP2190180.025mg
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

FZR1/CDH1 Monoclonal specifically detects FZR1/CDH1 in Human, Mouse samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZR1/CDH1
Applications ELISA, Western Blot
Classification Monoclonal
Clone AR38.2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000
Gene Alias anti-CDC20-like protein 1 anti-CDH1 anti-Cdh1/Hct1 homolog anti-Fizzy-related protein homolog anti-FYR anti-Fzr anti-FZR1 anti-hCDH1 CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), FYR, Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242, KIAA1242fizzy-related protein homolog
Gene Symbols FZR1
Host Species Mouse
Immunogen His-tagged recombinant FZR1/CDH1
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51343
Test Specificity NBP2-19018 recognizes Human and mouse.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.