Learn More
Memmert™ Full-Sight Glass Door
Product Code. 15745288 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
CTC256 Environmental Test Chamber
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15745288 CTC256 Environmental Test Chamber Each
1 options
Product Code. 15745288
Brand: Memmert™ CTC256B0
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Memmert™ Full-Sight Glass Door with 5-layer insulating glazing for use with Environmental Test Chambers.
  • 5-layer insulating glazing
  • Heated
Specifications
For Use With (Equipment) CTC256 Environmental Test Chamber
Material Glass
Type Full-Sight Door
Door Style Full-sight
Includes 5-layer insulating glazing