missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled 9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13865-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predomitly in brain, testis, eye, skeletal muscle, and kidney.Specifications
Frizzled 9 | |
Polyclonal | |
Unconjugated | |
FZD9 | |
CD349, Frizzled 9, Fz 9, FZD3, FZD9, FzE6, hFz9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14371, 266608, 8326 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00144, Q8K4C8, Q9R216 | |
Fzd9 | |
Frizzled 9 Fusion Protein Ag4847 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |