missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled 9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Frizzled 9 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892264
|
Proteintech
13865-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16882264
|
Proteintech
13865-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predomitly in brain, testis, eye, skeletal muscle, and kidney.Specifications
Frizzled 9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O00144, Q8K4C8, Q9R216 | |
14371, 266608, 8326 | |
Frizzled 9 Fusion Protein Ag4847 | |
Primary | |
-20°C | |
FZD9 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD349, Frizzled 9, Fz 9, FZD3, FZD9, FzE6, hFz9 | |
Fzd9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |