Learn More
H-B Instrument™ FRIO-Temp™ Calibrated Electronic Verification Lollipop Stem Thermometers
For use in digital verification of sample temperatures in general applications. H-B Instrument™ FRIO-Temp™ Calibrated Electronic Verification Lollipop Stem Thermometers render accurate temperature reading without sample contamination.
Brand:  H-B Instrument™ B61000-0700
Additional Details : Weight : 0.39460kg
 View more versions of this product
Product Code. 17560676
  • 955.00 NOK / Each
Estimated Shipment: 24-07-2024
to see stock.
Alert:
Not for use in ovens or freezers
Description
Description
  • Entire digital unit can be placed directly in refrigerators, incubators and ambient rooms
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized
    • Certifications
      ISO 17025, CE marked
Specifications
Specifications
Glass Beads
0°, 50°C
4.5 in.
2 in.
Stainless Steel
1.5 in.
70°C
0°C to 70°C
±0.5°C
30 mL Plastic Bottle
Digital verification of sample temperatures in general applications
114 mm
50 mm
0.1°C
Digital Thermometer
0°C
Yes
38 mm