Learn More
Karl Hecht™ Freezer Thermometer
Description
- Officially tested for accurate freezer temperature monitoring.
- Includes holder.
Specifications
Specifications
| Includes | Holder |
| Product Type | Thermometer |
| Temperature Range (Metric) | -35°C to +25°C |
| For Use With (Equipment) | Freezer |